Learn More
Promega Nuclei Lysis Solution for Wizard™ Genomic DNA Purification Kits
Component of Wizard, Wizard SV and Wizard SV 96 Genomic DNA Purification Systems, which are intended for isolating genomic DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria.
Supplier:  Promega A7941
Encompass
 Afficher plus de versions de ce produit
Numéro de catalogue. PRA7941
Il en reste null
Description
Description
  • Reagent used in DNA Purification Protocols
  • Component of Wizard(R), Wizard(R) SV and Wizard(R) SV 96 Genomic DNA Purification Systems available as a standalone item.
Spécifications
Identifiants chimiques
Wizard™ genomic DNA purification kit
Isolation of DNA from white Blood cells, tissue culture cells, animal tissue, plant tissue, yeast and gram positive and gram negative bacteria
Spécifications
15°C to 30°C
Isolation of DNA from white Blood cells, tissue culture cells, animal tissue, plant tissue, yeast and gram positive and gram negative bacteria
50 mL
Wizard™ genomic DNA purification kit
Nuclei Lysis Solution
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS