missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega pGL4 Luciferase Reporter Vectors
Available in numerous configurations for enhanced usability and convenience
Supplier: Promega E8471
Description
- Reporter Vectors Ready to Use in Your Signaling Pathway Without Cloning
- Offer a number of response elements that are involved in nuclear response, kinase and other signaling pathways
- Each vector included an optimized firefly luciferase gene that codes a reporter with a one-hour half-life
- Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line
Chemical Identifiers
| -30°C to -10°C | |
| 1 μg/μL | |
| 20 μg | |
| Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis |
Specifications
| Hygro | |
| hPEST | |
| 10mM Tris HCl pH 7.4, 1mM EDTA | |
| 1 μg/μL | |
| 20 μg | |
| Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis |
| CRE | |
| luc2A | |
| -30°C to -10°C | |
| RV Primer 3 and RV Primer 4 | |
| pGL4.29[luc2P/CRE/Hygro] |