missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega MagneSphere™ Technology Magnetic Separation Stand
Use in conjunction with the PolyATtract Systems. Ideal for applications requiring multiple paramagnetic isolations of biomolecules.
$105.41 - $362.35
Specifications
| Content And Storage | 15°C to 30°C |
|---|---|
| For Use With (Equipment) | MagneSphere™ and PolyATtract™ system components |
| For Use With (Application) | Ideal for applications requiring multiple paramagnetic isolations of biomolecules |
| Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
PRZ5332
|
Promega
Z5332 |
1.5 mL | 2-Position |
Each for $105.41
|
|
||||
PRZ5342
|
Promega
Z5342 |
1.5 mL | 12-Position |
Each for $362.35
|
|
||||
PRZ5331
|
Promega
Z5331 |
0.5 mL | 2-Position |
Each for $105.41
|
|
||||
PRZ5341
|
Promega
Z5341 |
0.5 mL | 12-Position |
Each for $355.75
|
|
||||
PRZ5333
|
Promega
Z5333 |
12 x 75 mm | 2-Position |
Each for $106.72
|
|
||||
PRZ5343
|
Promega
Z5343 |
12 x 75 mm | 12-Position |
Each for $362.35
|
|
||||
Description
- Use with both MagneSphere(R) and PolyATtract(R) System components
- Conduct simultaneous processing of the contents of two tubes or twelve tubes of various sizes
Specifications
| 15°C to 30°C | |
| MagneSphere™ and PolyATtract™ system components | |
| Ideal for applications requiring multiple paramagnetic isolations of biomolecules |