Learn More
Promega MagneSphere™ Technology Magnetic Separation Stand
Use in conjunction with the PolyATtract Systems. Ideal for applications requiring multiple paramagnetic isolations of biomolecules.
Supplier:  Promega Z5331
Encompass
 View more versions of this product
Catalog No. PRZ5331
Only null left
Description
Description
  • Use with both MagneSphere(R) and PolyATtract(R) System components
  • Conduct simultaneous processing of the contents of two tubes or twelve tubes of various sizes
Specifications
Chemical Identifiers
15°C to 30°C
MagneSphere™ and PolyATtract™ system components
Ideal for applications requiring multiple paramagnetic isolations of biomolecules
Specifications
15°C to 30°C
Ideal for applications requiring multiple paramagnetic isolations of biomolecules
0.5 mL
MagneSphere™ and PolyATtract™ system components
2-Position
Product Suggestions
Product Suggestions