Learn More
Promega MagneSphere™ Technology Magnetic Separation Stand
Use in conjunction with the PolyATtract Systems. Ideal for applications requiring multiple paramagnetic isolations of biomolecules.
Supplier: Promega Z5343
Description
- Use with both MagneSphere(R) and PolyATtract(R) System components
- Conduct simultaneous processing of the contents of two tubes or twelve tubes of various sizes
Chemical Identifiers
| 15°C to 30°C | |
| MagneSphere™ and PolyATtract™ system components | |
| Ideal for applications requiring multiple paramagnetic isolations of biomolecules |
Specifications
| 15°C to 30°C | |
| Ideal for applications requiring multiple paramagnetic isolations of biomolecules | |
| 12 x 75 mm |
| MagneSphere™ and PolyATtract™ system components | |
| 12-Position |