Learn More
Promega MagaZorb™ DNA Miniprep Kits
The MagaZorb DNA Kit provides an easy, fast and cost-effective technique for isolating PCR-quality DNA.
Supplier: Promega MB1004
Description
- Fast Isolation of PCR-quality DNA from Various Samples
- All reagents provided, no reagent preparation required
- Magnetic purification method increases throughput and reproducibility
- No organic solvents
Identifiants chimiques
| 15°C to 30°C | |
| Isolating PCR-quality DNA | |
| Kit |
Spécifications
| 15°C to 30°C | |
| 4mL Proteinase K solution, 40mL Lysis Buffer, 100mL Binding Buffer, 4mL MagaZorb™ reagent, 2 x 200mL Wash Buffer, 120mL Elution Buffer | |
| Kit |
| Isolating PCR-quality DNA | |
| 200 Preps |
U.S. Patents 6,855,499 and other patents pending.