missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega MagaZorb™ DNA Miniprep Kits
Description
- Fast Isolation of PCR-quality DNA from Various Samples
- All reagents provided, no reagent preparation required
- Magnetic purification method increases throughput and reproducibility
- No organic solvents
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| For Use With (Application) | Isolating PCR-quality DNA |
| Includes | 4mL Proteinase K solution, 40mL Lysis Buffer, 100mL Binding Buffer, 4mL MagaZorb™ reagent, 2 x 200mL Wash Buffer, 120mL Elution Buffer |
| No. of Reactions | 200 Preps |
| Product Type | Kit |
U.S. Patents 6,855,499 and other patents pending.