missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GoScript™ Reverse Transcription System
Description
Comprend
GoScript Reverse Transcriptase, GoScript 5X Reaction Buffer, MgCl2, Oligo(dT)15 Primer, PCR Nucleotide Mix, Random Primers, Recombinant RNasin Ribonuclease Inhibitor, Nuclease-free water
- Reliable cDNA Synthesis for Sensitive Detection of Expression
- Available as a standalone enzyme, a complete reverse transcription kit or as a master mix with Oligo(dT) or Random Primers. RNasin(R) RNase Inhibitor protection included with Kit and Mixes
- Sensitive transcription of both high-copy and low-copy messages
- Transcribes short and long transcripts; processes through secondary structure
Spécifications
Spécifications
| Content And Storage | • MgCl2, 1 x 750 μL • GoScript™ 5X Reaction Buffer, 1 x 300 μL • GoScript™ Reverse Transcriptase, 1 x 50 μL • oligo(dT)15 primer, 1 x 50 μg • Nuclease-free Water, 1 x 200μL Store at –30° to –10°C. |
| For Use With (Application) | RT-PCR, Real Time PCR (qPCR) |
| No. of Reactions | 50 Reactions |
- U.S. Patent 6,242,235, Australian Patent 761757, Canadian Patent 2,335,153 and other patents and patents pending.
- U.S. Patent 5,552,302, Australian Patent 646803 and other patents.