Learn More
Promega ADP-Glo™ Max ADP Detection Assay Kit
Catalog No. PRV7002
Encompass
Change view
Click to view available options
Quantity:
10,000 Assays
Catalog No. Quantity
PRV7002 10,000 Assays
1 options
Catalog No. PRV7002 Supplier Promega Supplier No. V7002
The ADP-Glo™ Max Assay is a luminescent ADP detection assay that provides a universal, homogeneous, high-throughput screening method to measure ATPase or kinase activity by quantifying the amount of ADP produced in a reaction.
  • Luminescent Method to Measure ATPase or Kinase Activity
  • Screen library compounds for effects on target enzymes
  • Perform mode of action studies on enzymes of interest
  • Measure activity of immunoprecipitated ATPases/kinases
  • Use when higher ATP concentration is required (up to 5mM)
Specifications
Content And Storage -30°C to -10°C
Quantity 10,000 Assays