missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 300HL Rounded Cap Knee Pads

Catalog No. 19820276
Change view
Click to view available options
rounded cap knee pads
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820276 rounded cap knee pads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820276 Supplier Ergodyne™ Supplier No. 18301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Short, hard cap is best for swiveling and pivoting.

Specifications

Color White Cap
For Use With (Application) Construction, Masonry, Freight/Baggage,Flooring, Maintenance, Utilities, Carpentry
Material 840D Nylon