missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ ProFlex™ 925F(x) Standard Dorsal Impact-Reducing Gloves
Description
- Full TPR armor with technical foam padding
- Hi-Vis AX Suede™ with non-slip dot gripping palm
- Armortex™-reinforced thumb saddle and index finger
- Hi-vis breathable mesh construction
- Padded neoprene cuff with reflective binding
- Pull-on tab with ID space to write user's name
Spécifications
Spécifications
| Cuff Material | Neoprene |
| Color | Hi-vis Lime Green, Black |
| Hand | Specific |
| Material | Thermoplastic Rubber |
| Product Type | Impact-Reducing Gloves |
| Size | Small |
| For Use With (Application) | Oil/Gas Drilling, Mining, and Heavy Construction |
| Glove Cuff Style | Open |
| Series | ProFlex™ 925F(x) |