missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 344 Short Cap Injected Gel Knee Pads with Comfort Straps

Catalog No. 19820285
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820285 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820285 Supplier Ergodyne™ Supplier No. 18444

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProFlex™ 344 Short Cap Injected Gel Knee Pads feature an innovative comfort strap system.

  • Short cap design for mobility when kneeling
  • Non-slip rubber cap grips kneeling surface
  • Injected gel and PU foam for all-day comfort
  • Padded straps on thigh and calf will not bind up
  • Plastic clips cinch tight and keep straps secure

Specifications

Color Black
Design Short Cap
For Use With (Application) Construction, Maintenance, Flooring, Mechanics