Learn More
Synbiosis™ ProcScan 3 Instrument Validation Procedures
GREENER_CHOICE
ProcScan 3 Instrument Validation Procedures take the user through each step in the verification of a Synbiosis™ system regulatory compliances and standard operational procedures (SOP). These simple procedures allow the user to verify that a system meets defined installation, operational, and performance quality specifications.
$7,909.66 - $9,563.89
Spécifications
For Use With (Equipment) ProcScan Instruments
For Use With (Application) Instrument Validation
voir les spécifications
Produits 3
Numéro de catalogue Numéro du manufacturier. Service Offering Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Service Offering Prix Quantity & Availability  
01257331
GREENER_CHOICE
Afficher les documents Détails de la promotion
Synbiosis™
OQPSCAN
Operational Qualification
chaque for $7,909.66
Il en reste null
01257332
GREENER_CHOICE
Afficher les documents Détails de la promotion
Synbiosis™
IQOQPSCAN
Installation and Operational Qualification
chaque for $8,099.13
Il en reste null
01257333
GREENER_CHOICE
Afficher les documents Détails de la promotion
Synbiosis™
IQOQPQPSCAN
Installation, Operational, and Performance Qualification
chaque for $9,563.89
Il en reste null
Description
Description
  • Installation Qualification (IQ): Verifies that when a system is unpacked it conforms with all purchasing, shipping, and unpacking documents. Once this procedure has been followed the system is then ready for the operational check process.
  • Operational Qualification (OQ): Establishes if the system meets all the operation and functionality specifications. An operational test is also included at this stage.
  • Performance Qualification (PQ): Establishes if the system meets the specification for the nature of the purpose it was intended for. This is verified by using a simple target to establish if the system can meet performance criteria.
  • Documentation: Each set of IQ/OQ/PQ documents contains a complete set of procedures and checklists for following the verification steps. Included with each pack is a simple calibration device (target image) so that the performance of the system can be checked against the PQ requirements. Each set of documents is unique to a particular system and has to be ordered with the system itself. The document pack has full instructions for use and implementation.
Spécifications
Spécifications
ProcScan Instruments
Instrument Validation
Videos
FDS