Learn More
Invitrogen™ ProcartaPlex Human Basic Kit
Description
Comprend
- Reading Buffer
- Wash Buffer 10x
- Streptavidin-PE
- Universal Assay Buffer
- Detection Antibody Diluent Human
- PCR 8-Tube Strip
- Adhesive Film
- Greiner Microplate black 96 well
ProcartaPlex™ immunoassays are our ideal solution for assessing multiple protein biomarkers in a single sample.
Our ProcartaPlex assays use Luminex ™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins.
ProcartaPlex™ assays are subject to the same stringent and robust development and manufacturing as are our extensively validated Platinum and Instant ELISA kits. Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma. Explore our broad portfolio of pre-configures panels and combinable single analytes. For combination of only Simplex Kits an additional Basic Kit is required. This kit includes all non-target specific reagents - assay diluent, wash buffers, SA-PE and plates.
This kit does not contain a Multiscreen Filter Plate.
Suitable Sample Types
Cell culture supernatant, serum, plasma
Sample Volume
Serum, Plasma: 25 μL; CCS: 50 μL
Reported Application:
Multiplex Immunoassay
Spécifications
Spécifications
| Product Type | Basic and Setup Kits |
| Quantity | 96 Tests |
| Species | Human |
| Combinability | Combinable |
| Assay Sensitivity | See Certificate of Analysis |
| For Use With (Equipment) | Luminex™ Instruments |
| Product Line | ProcartaPlex |
For Research Use Only. Not for use in diagnostic procedures.