missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Primary Rat Cortex Neurons

Reliable rat cortical neurons in a convenient cryopreserved format provide an ideal source of neurons to enable neuroscience cell-based assays.

$548.00 - $1,298.00

Spécifications

Content And Storage Store in liquid nitrogen, vapor-phase.
Age Fetal
Cell Type Neural Cells
Culture Type Mammalian Cell Culture
Form Cryopreserved
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
A1084001
Encompass
Afficher les documents Détails de la promotion
Gibco™
A1084001
1 x 106 cells
chaque for $548.00
Il en reste null
 
A1084002
Encompass
Afficher les documents Détails de la promotion
Gibco™
A1084002
4 x 106
chaque for $1,298.00
Il en reste null
 
Description

Description

Gibco Primary Rat Cortex Neurons (RCNs) are isolated and cryopreserved from day-18 Fischer 344 rat embryos. These cryopreserved RCNs offer a flexible, ready-to-use, and high-quality source of cells for neuronal experiments. They consistently yield post-thaw viability in the range of 50–80% and maintain high neuronal purity. Cryopreserved neurons are suitable for various applications requiring primary rat brain neurons, including patch-clamp electrophysiology, MEA, neurite outgrowth, synaptic protein expression, and neuron toxicity assays.

  • Convenience—these cryopreserved rat cortical neurons (RCNs) can be stored until needed and are ready to use when you plan your cell-based assays. They provide a convenient alternative to isolating neurons on your own.
  • Reliability—harvested from the cortex, these cryopreserved neurons meet rigorous quality standards to help ensure consistent performance
  • High Quality—every batch of Gibco Rat Cortex Neurons undergoes thorough testing, including viability assessment, immunofluorescence staining to confirm neuronal purity, sterility testing, and mycoplasma testing
  • Availability—our large-scale manufacturing capabilities allow us to support recurring orders of significant size and other requests, helping ensure you have the cells you need to complete your research studies

Culture Gibco Primary Rat Cortex Neurons with the B-27 Plus Neuronal Culture System to achieve the most robust performance.

Spécifications

Spécifications

Store in liquid nitrogen, vapor-phase.
Neural Cells
Cryopreserved
Cortex
Rat Cortex Neuron
Fetal
Mammalian Cell Culture
Rat
Normal
Sterility, Viability, Mycoplasma
Videos
FDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.