missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Prepared Microscope Slide - Ascaris and Onion Mitosis

Numéro de catalogue. S29649
Modifier l'affichage
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
S29649 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. S29649 Fournisseur Eisco™ Code fournisseur BS18034
Il en reste null

Prepared microscope slide of mitosis in ascaris and onion (allium) plants

  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization
  • Ideal for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows all stages of mitosis in animal and plant cells

Spécifications

Biosafety Level 1
Concentration 10%
Material Glass
Type Microbiology Slide
For Use With (Application) Microscopy
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Quantity 1
Species Ascaris and Onion
Size 1 x 3 in.
Slide Type Prepared Slide
Afficher plus de résultats Afficher moins de résultats