Learn More
Fisherbrand™ Pre-Filled Bead Mill Tubes
GREENER_CHOICE
For use with Bead Mill 4 and Bead Mill 24 homogenizers. May also be used with most other commercial Bead Mills.
$413.97 - $524.35
Products 7
Catalog Number Mfr. No. Product Type Capacity (Metric) Particle Size Material Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Capacity (Metric) Particle Size Material Price Quantity & Availability  
15340152
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
15340152
Tough Microorganism Lysing Mix Tubes 2 mL 0.5mm Glass
Pack of 50 Each for $413.97
Only null left
15340153
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
15340153
Soft Tissue Homogenizing Mix Tubes 2 mL 1.4 mm Ceramic
Pack of 50 Each for $413.97
Only null left
15340151
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
15340151
Hard Tissue Grinding Mix Tubes 2 mL 2.4 mm Metal
Pack of 50 Each for $413.97
Only null left
15340154
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
15340154
Hard Tissue Homogenizing Mix Tubes 2 mL 2.8 mm Ceramic
Pack of 50 Each for $413.97
Only null left
15340156
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
15340156
Soft Tissue Homogenizing Mix Tubes 7 mL 1.4 mm Ceramic
Pack of 50 Each for $524.35
Only null left
15340155
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
15340155
Hard Tissue Grinding Mix Tubes 7 mL 2.4 mm Metal
Pack of 50 Each for $524.35
Only null left
15340157
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
15340157
Hard Tissue Grinding Mix Tubes 7 mL 2.8 mm Ceramic
Pack of 50 Each for $524.35
Only null left
Description
Description
  • Polypropylene tubes closed with screw caps
  • Non-Sterile