missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Pre-Filled Bead Mill Tubes
GREENER_CHOICE

For use with Bead Mill 4 and Bead Mill 24 homogenizers. May also be used with most other commercial Bead Mills.

Supplier:  Fisherbrand™ 15340153

 Afficher plus de versions de ce produit

Numéro de catalogue. 15340153


Il en reste null

Comprend: 50 2mL tubes prefilled with 1.4mm ceramic beads

Description

Description

  • Used for processing brain, liver, kidney, lung, spleen
  • 2mL tubes closed with screw cover and o-ring
  • RNase/DNase free
  • Processing range: 300μL to 1.8mL
  • Non-Sterile
Spécifications

Spécifications

Soft Tissue Homogenizing Mix Tubes
1.4 mm
10.8 mm
Ceramic
Bead Mill 4 and 24 Homogenizers
2 mL
300 μL to 1.8 mL
46 mm (without cap); 49 mm (with cap)
Yes
Yes
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions