missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ DRIFIRE™ POWER GRID™ Fleece FR Balaclava

Catalog No. 17001746
Encompass
Change view
Click to view available options
Arc Flash Rating:
15 cal/cm²
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash Rating
17001746 15 cal/cm²
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17001746 Supplier National Safety Apparel™ Supplier No. H92FWPS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides warmth, protection and comfort. Flame resistant and adjusts to cover everything except eyes.

  • Versatile Wear Options: Designed to be worn multiple ways for flexible coverage and comfort
  • Long-Lasting FR Protection: Inherently flame-resistant, with protection that won’t wash or wear out
  • Seamless Comfort: Flat lock seams prevent chafing and add comfort for all-day wear
  • Flexible Four-Way Stretch: Moves with you, providing a secure and comfortable fit
  • Advanced Breathability: Grid fabric technology promotes airflow to prevent overheating
  • Moisture-Wicking Power: High-performance moisture management keeps you dry, even during intense activity
  • Lightweight Warmth: Delivers warmth without the bulk, perfect for cold weather work
  • Made in the USA

Specifications

Product Type FR Balaclava
Size Universal
Material Rayon, Modacrylic, Para-aramid, Spandex
Color Black
Closure Type None
Disposable Reusable
Seam Type Stitched
Arc Flash Rating 15 cal/cm²
Certifications/Compliance CAT 2, NFPA 70E, NFPA 2112, ASTM F1506, CSA Z462, OSHA 1910.269
Includes Balaclava
Construction Balaclava
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Resistance Flame
Hood Type Attached
Style Balaclava
Show More Show Less