Learn More
Alconox™ Powdered Precision Cleaner
Concentrated, anionic detergent for manual and ultrasonic cleaning
Supplier: Alconox™ 1125
Description
Concentrated, anionic detergent for manual and ultrasonic cleaning
Alconox consists primarily of a homogeneous blend of sodium linear alkylaryl sulfonate, alcohol sulfate, phosphates, carbonates.
- Biodegradable and readily disposable
- Penetrating wetting power to save time
- Replaces corrosive acids and hazardous solvents
- Free rinsing gives reliable results without interfering residues
- Use to pass cleaning validation tests for lab accreditation and plant inspection approval
- Dilute 1:100 for typical use (10g in 1L of water)
- USDA authorized for use in federally inspected meat and poultry plants
Used to clean:
Healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus, catheters, tubing, pipes, radioactive contaminated articles, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal castings, forgings and stampings, industrial parts, tanks and reactors, passes inhibitory residue test for water analysis.
Used to remove:
Soil, grit, grime, buffing compound, slime, grease, oils, blood, tissue, salts, deposits, particulates, solvents, chemicals, radioisotopes, radioactive contaminations, silicon oils, mold release agents.
Surfaces cleaned:
Corrosion inhibited formulation recommended for glass, metal,stainless steel, porcelain, ceramic, plastic, rubber and fiberglass, can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly.
Cleaning method:
Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace. Will foam—not for spray or machine use.
Shelf life 2 years from date of manufacture.
Order Info
0.5kg (1 lb.) makes 49.2L (13 gal.) cleaning solution.
Identifiants chimiques
| Powdered Precision Cleaner | |
| 9.5 | |
| Store closed and upright in a cool dry place at 15° to 30°C. |
| Powder | |
| White, Cream | |
| Cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass |
Spécifications
| Powdered Precision Cleaner | |
| 9.5 | |
| Carton | |
| Cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass | |
| 11.3 kg |
| Powder | |
| White, Cream | |
| Store closed and upright in a cool dry place at 15° to 30°C. | |
| 25 lb. |