missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC ORGK82Z
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| K | 1000μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Potassium Oil Standard | |
| 7440-09-7 , 8042-47-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.5,99.5 | |
| MFCD00133776 | |
| Certified Reference Material | |
| [K] | |
| potassium | |
| 39.10 | |
| 5462222 | |
| Oil/Plasma |
| Triple-leached LDPE bottle | |
| With Cerfiticate of Analysis | |
| 0.5,99.5 | |
| K | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| ZLMJMSJWJFRBEC-UHFFFAOYSA-N | |
| Base Oil 75 | |
| Petrochemical Testing | |
| 50 g | |
| CHEBI:26216 | |
| Matrix Concentration Cat. No. Base Oil 1000μg/g ORG-K8-2Z Base Oil 5000μg/g ORG-K8-4Z |
Chemical Identifiers
| 7440-09-7 | |
| 39.10 | |
| ZLMJMSJWJFRBEC-UHFFFAOYSA-N | |
| CHEBI:26216 | |
| [K] |
| K | |
| MFCD00133776 | |
| 5462222 | |
| potassium |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only