accessibility menu, dialog, popup

UserName

K

CAS RN 7440-09-7

IUPAC Name: potassium
Synonyms: potassium|kalium|potasio|potassium atom|humic acid
Molecular Weight (g/mol): 39.1
Molecular Formula: K
InChi Key: ZLMJMSJWJFRBEC-UHFFFAOYSA-N
SMILES: [K]
Grade 
  • (1)
  • (1)
  • (2)
Solvent or Matrix 
  • (1)
  • (2)
Industry Type 
  • (2)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)