missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, 15% (w/v), Stabilized, Ricca Chemical

Catalog No. 630516
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Poly Bottle
Black Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
630516 500 mL Amber Poly Bottle
630532 1 L Amber Poly Bottle
63051 4 L Black Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 630516 Supplier Ricca Chemical Company Supplier No. 630516

Chemical Identifiers

CAS 7681-11-0 , 7732-18-5 , 144-55-8
Molecular Formula IK
Molecular Weight (g/mol) 166.00
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium iodide
SMILES [K+].[I-]

Specifications

Color Colorless to Yellow
CAS Min % 12.79, 85.0, 0.01
CAS Max % 13.31, 89.0, 0.01
Percent Purity 15%
Grade Laboratory
Packaging Amber Poly Bottle
Treatment(s) Stabilized
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 15% (w/v)
Chemical Name or Material Potassium Iodide
Show More Show Less