missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, 10% (w/v), Stabilized, Ricca Chemical

Supplier:  Ricca Chemical Company RI63005

 View more versions of this product

Catalog No. 6300-5


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Iodide
7681-11-0 , 7732-18-5
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
[K+].[I-]
166.003
CHEBI:8346
Laboratory
Stabilized
20 L
10% (w/v)
Colorless
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
10%
Cubitainer
Aqueous Solution
Liquid

Chemical Identifiers

7681-11-0
166.003
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
CHEBI:8346
[K+].[I-]
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions