missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, 10% (w/v), Stabilized, Ricca Chemical

Catalog No. p-3252182
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Poly Bottle
Black Poly Bottle
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
630016 500 mL Amber Poly Bottle
630032 1 L Amber Poly Bottle
63001 4 L Black Poly Bottle
63005 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 630016 Supplier Ricca Chemical Company Supplier No. 630016
Only null left

Chemical Identifiers

CAS 7681-11-0 , 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Color Colorless
Percent Purity 10%
Grade Laboratory
Packaging Amber Poly Bottle
Treatment(s) Stabilized
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Potassium Iodide