missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate-Iodide, 0.100 N (N/10), Ricca Chemical
$285.67 - $946.10
Specifications
| Chemical Name or Material | Potassium Iodate-Iodide |
|---|---|
| CAS | 7732-18-5 |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Grade | Laboratory |
| pH | Slightly Alkaline |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
628516
|
Ricca Chemical Company
628516 |
500 mL | Amber Glass Bottle |
Each for $285.67
|
|
||||
|
6285-32
|
Ricca Chemical Company
628532 |
1 L | Amber Glass Bottle |
Each for $375.00
|
|
||||
|
6285-1
|
Ricca Chemical Company
62851 |
4 L | Amber Glass Bottle |
Each for $558.70
|
|
||||
|
6285-5
|
Ricca Chemical Company
62855 |
20 L | Cubitainer |
Each for $946.10
|
|
||||
Specifications
| Potassium Iodate-Iodide | |
| dihydrogen oxide, dihydrogen monoxide | |
| Slightly Alkaline |
| 7732-18-5 | |
| Laboratory |