missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate-Iodide, 0.100 N (N/10), Ricca Chemical
Supplier: Ricca Chemical Company 62851
Specifications
| Potassium Iodate-Iodide | |
| dihydrogen oxide, dihydrogen monoxide | |
| 0.100N (N/10) | |
| Slightly Alkaline |
| 7732-18-5 | |
| Laboratory | |
| Amber Glass Bottle | |
| 4 L |