missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate-Iodide, 0.100 N (N/10), Ricca Chemical

Supplier:  Ricca Chemical Company 628532

 View more versions of this product

Catalog No. 6285-32

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Iodate-Iodide
dihydrogen oxide, dihydrogen monoxide
0.100N (N/10)
Slightly Alkaline
7732-18-5
Laboratory
Amber Glass Bottle
1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions