missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate, 0.100 N (N/10), 0.01667 M (M/60), Ricca Chemical
$165.77 - $585.00
Chemical Identifiers
| CAS | 7758-05-6 |
|---|---|
| Molecular Formula | IKO3 |
| Molecular Weight (g/mol) | 214.00 |
| InChI Key | JLKDVMWYMMLWTI-UHFFFAOYSA-M |
| Synonym | potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat |
| PubChem CID | 23665710 |
| IUPAC Name | potassium;iodate |
| SMILES | [O-]I(=O)=O.[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
627516
|
Ricca Chemical Company
627516 |
500 mL | Amber Glass Bottle |
Each for $165.77
|
|
||||
|
6275-32
|
Ricca Chemical Company
627532 |
1 L | Amber Glass Bottle |
Each for $310.20
|
|
||||
|
6275-1
|
Ricca Chemical Company
RI62751 |
4 L | Amber Glass Bottle |
Each for $585.00
|
|
||||
Chemical Identifiers
| 7758-05-6 | |
| 214.00 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| potassium;iodate |
| IKO3 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| 23665710 | |
| [O-]I(=O)=O.[K+] |
Specifications
| Colorless | |
| 98.0,0.36 | |
| IKO3 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| potassium;iodate | |
| 23665710 | |
| Amber Glass Bottle | |
| Liquid | |
| Potassium Iodate |
| 7758-05-6 , 7732-18-5 | |
| 100.0,0.37 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| [O-]I(=O)=O.[K+] | |
| 214.00 | |
| Laboratory | |
| 500 mL | |
| 0.100N,0.01667M |