missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate, 0.100 N (N/10), 0.01667 M (M/60), Ricca Chemical
Supplier: Ricca Chemical Company RI62751
Specifications
| Potassium Iodate | |
| 7758-05-6 , 7732-18-5 | |
| 100.0,0.37 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| [O-]I(=O)=O.[K+] | |
| 214.00 | |
| Laboratory | |
| 4 L | |
| 0.100N,0.01667M |
| Colorless | |
| 98.0,0.36 | |
| IKO3 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| potassium;iodate | |
| 23665710 | |
| Amber Glass Bottle | |
| Liquid |
Chemical Identifiers
| 7758-05-6 | |
| 214.00 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| potassium;iodate |
| IKO3 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| 23665710 | |
| [O-]I(=O)=O.[K+] |