missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Solution, (Nominal 12.8 mS/cm), Certipur™, MilliporeSigma™,
Description
Recommended Storage
+15°C to +25°C.
Spécifications
Spécifications
| Density | 1.003 g/3 |
| Packaging | Plastic Bottle |
| Quantity | 500 mL |
Sécurité et manipulation
| Recommended Storage | +15°C to +25°C. |