missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
$121.75 - $906.25
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| IUPAC Name | potassium chloride |
| SMILES | [Cl-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R5888141500
|
Ricca Chemical Company
R5888141500A |
500 mL |
Each for $121.75
|
|
|||||
|
R58881411A
|
Ricca Chemical Company
R58881411A |
1 L |
Each for $159.28
|
|
|||||
|
R58881414A
|
Ricca Chemical Company
R58881414A |
4 L |
Each for $281.19
|
|
|||||
|
R588814110F
|
Ricca Chemical Company
R588814110F |
10 L |
Each for $534.25
|
|
|||||
|
R588814120F
|
Ricca Chemical Company
R588814120F |
20 L |
Each for $906.25
|
|
|||||
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| CHEBI:32588 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| 4873 | |
| potassium chloride |
Specifications
| Natural Poly Bottle | |
| 15.0,85.0 | |
| ClK | |
| Colorless | |
| 100°C | |
| Conductivity Standard | |
| [Cl-].[K+] | |
| 74.55 | |
| 4873 | |
| Potassium Chloride Conductivity Standard, 1, 413 μmho/cm at 25°C |
| 7447-40-7,7732-18-5 | |
| Liquid | |
| MFCD00011360 | |
| Approximately 7 | |
| Conductivity Standard | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 500 mL | |
| CHEBI:32588 |
Safety and Handling
ShelfLife : 18 months