missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company R58881411A

 View more versions of this product

Catalog No. R58881411A


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Chloride Conductivity Standard
Natural Poly Bottle
15.0,85.0
ClK
Colorless
100°C
Conductivity Standard
[Cl-].[K+]
74.55
4873
1, 413 μmho/cm at 25°C
7447-40-7,7732-18-5
Liquid
MFCD00011360
Approximately 7
Conductivity Standard
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
1 L
CHEBI:32588

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
ClK
MFCD00011360
4873
potassium chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents

Promotions

Promotions