missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company R588814110F
Specifications
| Potassium Chloride Conductivity Standard | |
| Cubitainer | |
| 15.0,85.0 | |
| ClK | |
| Colorless | |
| 100°C | |
| Conductivity Standard | |
| [Cl-].[K+] | |
| 74.55 | |
| 4873 |
| 1, 413 μmho/cm at 25°C | |
| 7447-40-7,7732-18-5 | |
| Liquid | |
| MFCD00011360 | |
| Approximately 7 | |
| Conductivity Standard | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 10 L | |
| CHEBI:32588 |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| CHEBI:32588 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| 4873 | |
| potassium chloride |
Safety and Handling
ShelfLife : 18 months