missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, 0.159M, Conductivity Standard
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Assay Percent Range | 0.159M |
| Physical Form | Liquid |
| Color | Colorless |
| IUPAC Name | potassium chloride |
| Quantity | 500 mL |
| Formula Weight | 74.55 |
| Concentration or Composition (by Analyte or Components) | Potassium chloride: 1.2%; Water: 98.8% |
| Chemical Name or Material | Potassium chloride, 20, 000μmho at 25°C |
Sécurité et manipulation
| EINECSNumber | 231-211-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only