missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Antimonyl Tartrate, 0.2743% (w/v) (1.3715 g/500 mL), Ricca Chemical
$214.44 - $328.31
Chemical Identifiers
| CAS | 11071-15-1 |
|---|---|
| Molecular Formula | C8H4K2O12Sb2 |
| Molecular Weight (g/mol) | 613.83 |
| InChI Key | GUJUCWZGYWASLH-SOMOIXMJSA-J |
Chemical Identifiers
| 11071-15-1 | |
| C8H4K2O12Sb2 | |
| 613.83 | |
| GUJUCWZGYWASLH-SOMOIXMJSA-J |
Specifications
| Colorless | |
| C8H4K2O12Sb2 | |
| 613.83 | |
| Laboratory | |
| Aqueous Solution | |
| Liquid | |
| Potassium Antimonyl Tartrate |
| 11071-15-1,7732-18-5 | |
| GUJUCWZGYWASLH-SOMOIXMJSA-J | |
| 0.2743% | |
| Natural Poly Bottle | |
| 500 mL | |
| 0.2743% (w/v),1.3715g/500mL |