accessibility menu, dialog, popup

UserName

KSb(R)(R)OOOOOO222

CAS RN 11071-15-1

IUPAC Name: diantimony(3+) dipotassium bis((2R,3R)-2,3-dioxidobutanedioate)
Synonyms: diantimony(3+) dipotassium bis((2R,3R)-2,3-dioxidobutanedioate)
Molecular Weight (g/mol): 613.83
Molecular Formula: C8H4K2O12Sb2
InChi Key: GUJUCWZGYWASLH-SOMOIXMJSA-J
SMILES: [K+].[K+].[Sb+3].[Sb+3].[O-][C@H]([C@@H]([O-])C([O-])=O)C([O-])=O.[O-][C@H]([C@@H]([O-])C([O-])=O)C([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)