missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Antimonyl Tartrate, 0.2743% (w/v) (1.3715 g/500 mL), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| Percent Purity | 0.2743% |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| Quantity | 1 L |
| Physical Form | Liquid |
| Concentration or Composition Notes | 0.2743% (w/v),1.3715g/500mL |
| Chemical Name or Material | Potassium Antimonyl Tartrate |