missing translation for 'onlineSavingsMsg'
Learn More

Potassium Antimonyl Tartrate, 0.2743% (w/v) (1.3715 g/500 mL), Ricca Chemical

Catalog No. 587216
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
587216 500 mL Natural Poly Bottle
587232 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 587216 Supplier Ricca Chemical Company Supplier No. 587216

Chemical Identifiers

CAS 11071-15-1 , 7732-18-5
Molecular Formula C8H4K2O12Sb2
Molecular Weight (g/mol) 613.83
InChI Key GUJUCWZGYWASLH-SOMOIXMJSA-J

Specifications

Color Colorless
Percent Purity 0.2743%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.2743% (w/v),1.3715g/500mL
Chemical Name or Material Potassium Antimonyl Tartrate