Learn More
Qorpak™ Polystyrene Snap Cap Vial
Qorpak™ Polystyrene Snap Cap Vials offer great rigidity and glass-like clarity.
Supplier:  Qorpak™ 289606
 Afficher plus de versions de ce produit
Numéro de catalogue. 02993910
Il en reste null
Description
Description
  • Lightweight
  • Glass-like clarity
  • Ideal for powders and non-liquid materials
  • Caps snap on and off quickly and easily
  • Suitable for storing bench top accessories and field samples
Spécifications
Identifiants chimiques
White
Polystyrene
Non-sterile
No
HDPE
No
No
Clear
No
Single-use
Bulk
Snap Cap
Not Autoclavable
Crimp/Snap Top Vial
Spécifications
White
Clear
Polystyrene
Non-sterile
20 Dram
144
HDPE
144/Cs.
Not Autoclavable
No
46 mm
38 mm
No
Single-use
No
Bulk
Snap Cap
No
68 mm
Crimp/Snap Top Vial
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS