missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 40, NF, Spectrum™ Chemical
PO134, 9005-66-7
Supplier: Spectrum Chemical Mfg Cor PO134200LTBL
| Quantity | 200 L |
|---|---|
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical Polysorbate 40, NF is used to solubilize essential oils. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Identifiants chimiques
| 9005-66-7 | |
| 522.68 | |
| DVRFSZFBSOKHQW-UHFFFAOYNA-N | |
| CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | |
| MFCD00165345 | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
Spécifications
| Polysorbate 40 | |
| 2 | |
| 200 L | |
| 0.0025 | |
| 41-52 | |
| 0.03 | |
| DVRFSZFBSOKHQW-UHFFFAOYNA-N | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate | |
| NF |
| 9005-66-7 | |
| 1 | |
| 89-105 | |
| Amber Glass Bottle | |
| (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | |
| MFCD00165345 | |
| CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 522.68 |