missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 40, NF, Spectrum™ Chemical
PO134, 9005-66-7
$238.36 - $49,178.25
Chemical Identifiers
| CAS | 9005-66-7 |
|---|---|
| Molecular Formula | (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 |
| Molecular Weight (g/mol) | 522.68 |
| MDL Number | MFCD00165345 |
| InChI Key | DVRFSZFBSOKHQW-UHFFFAOYNA-N |
| IUPAC Name | 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
| SMILES | CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18605384
|
N/A | 25 mL | Amber Glass Bottle |
Each for $238.36
|
|
||||
|
18605380
|
N/A | 500 mL | Amber Glass Bottle |
Each for $675.36
|
|
||||
|
18605381
|
N/A | 4 L | Amber Glass Bottle |
Each for $2,345.82
|
|
||||
|
18605382
|
N/A | 20 L | Amber Glass Bottle |
Each for $6,316.00
|
|
||||
|
18605383
|
N/A | 200 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Polysorbate 40, NF is used to solubilize essential oils. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
| 9005-66-7 | |
| 522.68 | |
| DVRFSZFBSOKHQW-UHFFFAOYNA-N | |
| CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | |
| MFCD00165345 | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
Specifications
| 9005-66-7 | |
| 1 | |
| 0.0025 | |
| (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | |
| MFCD00165345 | |
| CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 522.68 | |
| Polysorbate 40 |
| 2 | |
| 89-105 | |
| 41-52 | |
| 0.03 | |
| DVRFSZFBSOKHQW-UHFFFAOYNA-N | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate | |
| NF |