missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Polypropylene Sealing Film

Catalog No. 03840314
Change view
Click to view available options
Material:
Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
03840314 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03840314 Supplier BRANDTECH™ BRAND™ Supplier No. 701393ES

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

General purpose sealing film for ELISA and short-term storage

  • Polypropylene film with good chemical resistance
  • Designed for use with polypropylene and polystyrene plates
  • Certified DNase- and RNase-free
  • Pack of 100 non-sterile sheets

Specifications

Color Clear
Solvent Or Alcohol Resistant DMSO Resistant
Material Polypropylene
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96
No. per Pack 100 sheets
For Use With (Application) short--term storage, incubation, ELISA
Format ANSI/SLAS
For Use With (Equipment) Microplates/Deepwell Plates
Pre-slit No
Product Type Film, Seal, or Applicator
Adhesive Acrylic
Certified RNase/DNase-free Yes
Pierceable No
Temperature Range (Metric) -40°C to +120°C
Well Shape All well shapes
Plate Compatibility 96-well Plate
Show More Show Less