missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Cleanroom Wipe
GREENER_CHOICE

Engineered to absorb oil and other tough to clean chemicals

Supplier:  Fisherbrand™ FSNTP711FB

 Afficher plus de versions de ce produit

Numéro de catalogue. 17444149


Il en reste null

Description

Description

  • Fisherbrand cleanroom wipes are made from 100% melt-blown polypropylene
  • They are engineered to absorb oil and other tough to clean chemicals, yet soft enough for wiping sensitive surfaces
  • NTP wipes are manufactured in a cleanroom environment and cleanroom packaged
  • They double-bagged for added cleanliness and clean room use
  • Fisherbrand Nonwoven, 100% Melt-blown Polypropylene, ISO Class 5-6 (100-1000)
  • Cleanroom Wipe Fisherbrand 70% IPA/30% DI water, Polypropylene, 7 x 11 in.
  • ISO Cleanroom Class 5-8 Compatible wipe
Spécifications

Identifiants chimiques

Cleanroom Wipe
27.94 cm
Not Autoclavable
1-ply
11 in.
Polypropylene
Single-use
Non-sterile

Spécifications

Cleanroom Wipe
17.78 cm
27.94 cm
Not Autoclavable
1-ply
7 in.
11 in.
Polypropylene
Single-use
Non-sterile
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions