missing translation for 'onlineSavingsMsg'
Learn More

Platinum, plasma standard solution, Specpure™ 1000μg/mL

CAS: 7440-06-4 | Pt | 195.08 g/mol

$208.92 - $208.92

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13827AD
View Documents
Thermo Scientific Chemicals
013827AD
50 mL
Each for $208.92
Only null left
 
Description

Description

Platinum, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
CHEBI:33400
[Pt]
Pt
MFCD00011179
23939
platinum

Specifications

7440-06-4 , 7647-01-0 , 7732-18-5
Liquid
MFCD00011179
Miscible with water
[Pt]
195.08
23939
Hydrochloric acid: 20.0%; Antimony: 0.1%; Water: 79.9%
23 hPa (17mm Hg) at 20°C
Pt
Colorless
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
50 mL
CHEBI:33400
Platinum
Videos
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 231-595-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only