CAS RN 7440-06-4
Platinum, plasma standard solution, Specpure™ 1000μg/mL
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 MDL Number: MFCD00011179 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N PubChem CID: 23939 ChEBI: CHEBI:33400 IUPAC Name: platinum SMILES: [Pt]
Platinum, 10% on carbon, dry
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 MDL Number: MFCD00011179 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N Synonym: black,platin,sponge,platinum, metal,platinum, elemental,platine,platino,metal,platin german,iv ion PubChem CID: 23939 ChEBI: CHEBI:33400 IUPAC Name: platinum SMILES: [Pt]
Platinum ICP Standard, 1000 ppm Pt in 3% HCl, Ricca Chemical
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 MDL Number: MFCD00011179 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N PubChem CID: 23939 ChEBI: CHEBI:33400 IUPAC Name: platinum SMILES: [Pt]
Platinum ICP Standard, 10,000 ppm Pt in 20% HCl, Ricca Chemical
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N PubChem CID: 23939 ChEBI: CHEBI:33400 IUPAC Name: platinum SMILES: [Pt]
Platinum Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 MDL Number: MFCD00011179 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N PubChem CID: 23939 ChEBI: CHEBI:33400 IUPAC Name: platinum SMILES: [Pt]
Platinum Standard For ICP/MS, SPEX CertiPrep™
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 MDL Number: MFCD00011179 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N PubChem CID: 23939 ChEBI: CHEBI:33400 IUPAC Name: platinum SMILES: [Pt]
5 Percent Platinum on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst, Spectrum™ Chemical
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 MDL Number: MFCD00011179 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N IUPAC Name: platinum SMILES: [Pt]
0.8 Percent Platinum - 0.3 Percent Molybdenum on Carbon Powder, Reduced, Water Wet, 25 microns, BASF Catalyst, Spectrum™ Chemical
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 MDL Number: MFCD00011179 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N IUPAC Name: platinum SMILES: [Pt]
Platinum standard solution, for AAS, 1 mg/ml Pt in 10-20% HCl
CAS: 7440-06-4 | Pt | 195.08 g/mol
5 Percent Platinum on Activated Carbon Powder, Reduced, Water Wet, 27 microns, BASF Catalyst, Spectrum™ Chemical
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 MDL Number: MFCD00011179 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N IUPAC Name: platinum SMILES: [Pt]
5 Percent Platinum on Activated Carbon Powder, Unreduced, Water Wet, 27 microns, Spectrum™ Chemical
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 MDL Number: MFCD00011179 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N IUPAC Name: platinum SMILES: [Pt]
0.8 Percent Platinum on Carbon Powder, Reduced, Water Wet, 25 microns, BASF Catalyst, Spectrum™ Chemical
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 MDL Number: MFCD00011179 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N IUPAC Name: platinum SMILES: [Pt]
5 Percent Platinum on Calcium Carbonate Powder, Unreduced, Dry, 3 microns, BASF Catalyst, Spectrum™ Chemical
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 MDL Number: MFCD00011179 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N IUPAC Name: platinum SMILES: [Pt]
- 1
- 2