Learn More
Platinum(IV) oxide monohydrate, Adams Catalyst, Pt 75% min
Description
Hydrogenation catalyst. Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groupsPlatinum(IV) oxide monohydrate is used as a catalyst in hydrogenation, hydrogenolysis, dehydrogenation, and oxidation reactions in organic synthesis. It also finds application in ceramic industry, inorganic chemistry and in chemical synthesis. It is utilized for the selective reduction of alkenes without reducing nitro group.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Hydrogenation catalyst
- Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groups
Spécifications
Spécifications
| Melting Point | 450°C |
| Color | Black to Brown |
| Physical Form | Powder |
| Quantity | 1 g |
| Linear Formula | PtO2·H2O |
| UN Number | UN1479 |
| Merck Index | 14,7527 |
| Solubility Information | Insoluble in water,acid,aqua regiaSoluble in water and caustic potash solution; Insoluble in alcohol,acid and aqua regia. |
| Formula Weight | 245.10 (227.09 Anhydrous) |
| Concentration or Composition (by Analyte or Components) | Pt 75% min. |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H271 |
| Hazard Statement | GHS H Statement H272 May intensify fire oxidizer. |
| Precautionary Statement | P210-P220-P221-P280-P283-P306+P360-P370+P378q-P371+P380+P375-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: I |
| EINECSNumber | 215-223-0 |
| RTECSNumber | TP2506020 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only