missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum ICP Standard, 1000 ppm Pt in 3% HCl, Ricca Chemical
$867.70 - $1501.58
Chemical Identifiers
| CAS | 7440-06-4 |
|---|---|
| Molecular Formula | Pt |
| Molecular Weight (g/mol) | 195.08 |
| MDL Number | MFCD00011179 |
| InChI Key | BASFCYQUMIYNBI-UHFFFAOYSA-N |
| PubChem CID | 23939 |
| ChEBI | CHEBI:33400 |
| IUPAC Name | platinum |
| SMILES | [Pt] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PPT1KH-100
|
Ricca Chemical Company
PPT1KH100 |
100 mL | Natural LDPE Bottle |
Each for $867.70
|
|
||||
|
PPT1KH-500
|
Ricca Chemical Company
PPT1KH500 |
500 mL | Natural Poly Bottle |
Each for $1,501.58
|
|
||||
Chemical Identifiers
| 7440-06-4 | |
| 195.08 | |
| BASFCYQUMIYNBI-UHFFFAOYSA-N | |
| CHEBI:33400 | |
| [Pt] |
| Pt | |
| MFCD00011179 | |
| 23939 | |
| platinum |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm Pt | |
| 97.0,4.56,0.1 | |
| Liquid | |
| MFCD00011179 | |
| Approximately 0°C | |
| BASFCYQUMIYNBI-UHFFFAOYSA-N | |
| platinum | |
| 100 mL | |
| CHEBI:33400 |