missing translation for 'onlineSavingsMsg'
Learn More

Platinum ICP Standard, 1000 ppm Pt in 3% HCl, Ricca Chemical

Catalog No. PPT1KH100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PPT1KH100 100 mL Natural LDPE Bottle
PPT1KH500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PPT1KH100 Supplier Ricca Chemical Company Supplier No. PPT1KH100

Chemical Identifiers

CAS 7440-06-4 , 7647-01-0 , 7732-18-5
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Pt
CAS Min % 93.0, 4.38, 0.09
CAS Max % 97.0, 4.56, 0.1
Equivalence 1 mL = 1 mg Pt
Physical Form Liquid
Color Yellow
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Platinum ICP Standard
Show More Show Less