missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Plastic Pulley Collection

A collection of pulleys for use in classroom demonstrations.

Supplier:  Eisco™ PH0280A

 Afficher plus de versions de ce produit

Numéro de catalogue. S99351


Il en reste null

Description

Description

  • High quality plastic pulleys mounted in steel frame
  • 2 hooks - 1 attached to each end of the steel mounting for hanging
  • Capable of holding up to 30 lb. of weight
Spécifications

Identifiants chimiques

Plastic, Metal
30 lb.
Classroom Demonstrations
2 Hooks

Spécifications

Single Pulley
30 lb.
Classroom Demonstrations
2 Hooks
8.9 cm
1.63 lb.
Plastic, Metal
2 in.
5 cm
3.5 in.
0.74 kg
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions